Learn More
Biomatik Corporation Recombinant Insulin Like Growth Factor 1 (IGF1), 100UG
Supplier:  Biomatik Corporation RPU42024
Encompass_Preferred
Recombinant Pig Insulin Like Growth Factor 1 (IGF1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Insulin Like Growth Factor 1 (IGF1). Target Synonyms: IGFI; IGF1A; IBP1; MGF; Somatomedin C; Mechano Growth Gactor. Accession Number: P16545. Gene ID: 397491. Expression Region: Gly49~Ala118. Tag Info: N-terminal GST Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5592