Learn More
Biomatik Corporation Recombinant Integrin Beta 3 (ITGb3), 100UG
Catalog No. 501505595
Encompass_Preferred
Recombinant Human Integrin Beta 3 (ITGb3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Integrin Beta 3 (ITGb3). Target Synonyms: CD61; ITG-B3; GP3A; GP3-A; GpIIIa; Platelet Membrane Glycoprotein IIIa. Accession Number: P05106. Gene ID: 3690. Expression Region: Asp135~Ile377. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5595 Supplier Biomatik Corporation Supplier No. RPU42085
Recombinant Human Integrin Beta 3 (ITGb3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Integrin Beta 3 (ITGb3). Target Synonyms: CD61; ITG-B3; GP3A; GP3-A; GpIIIa; Platelet Membrane Glycoprotein IIIa. Accession Number: P05106. Gene ID: 3690. Expression Region: Asp135~Ile377. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.