Learn More
Biomatik Corporation Recombinant Intelectin 1 (ITLN1), 100UG
Supplier:  Biomatik Corporation RPU42090
Encompass_Preferred
Recombinant Human Intelectin 1 (ITLN1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Intelectin 1 (ITLN1). Target Synonyms: HL1; INTL; ITLN; LFR; HIntL; Omentin; Intelectin 1,Galactofuranose Binding; Intestinal Lactoferrin Receptor; Endothelial lectin HL-1. Accession Number: Q8WWA0. Gene ID: 55600. Expression Region: Cys31~Gly253. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5597