Learn More
Biomatik Corporation Recombinant Human Transcriptional enhancer factor TEF-5 (TEAD3), partial, 100UG
Supplier:  Biomatik Corporation RPC20167
Encompass_Preferred
Recombinant Human Transcriptional enhancer factor TEF-5 (TEAD3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TEAD3. Target Synonyms: DTEF-1TEA domain family member 3; TEAD-3. Accession Number: Q99594. Expression Region: 112~435aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 52.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4636