Learn More
Biomatik Corporation Recombinant Interferon Beta (IFNb), 100UG
Supplier:  Biomatik Corporation RPU42129
Encompass_Preferred
Recombinant Dog Interferon Beta (IFNb) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Dog (Canis familiaris; Canine). Target Name: Interferon Beta (IFNb). Target Synonyms: IFNB1; IFN-B; IFB; IFF; IFNB; Interferon Beta 1 Fibroblast. Accession Number: B6E116. Gene ID: 481558. Expression Region: Ala21~Asn186. Tag Info: N-terminal His and GST Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5601