Learn More
Biomatik Corporation Recombinant Interferon Gamma (IFNg), 100UG
Catalog No. 501505605
Encompass_Preferred
Recombinant Chicken Interferon Gamma (IFNg) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Chicken (Gallus). Target Name: Interferon Gamma (IFNg). Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Accession Number: P49708. Gene ID: 396054. Expression Region: His20~Cys164. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5605 Supplier Biomatik Corporation Supplier No. RPU42139
Recombinant Chicken Interferon Gamma (IFNg) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Chicken (Gallus). Target Name: Interferon Gamma (IFNg). Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Accession Number: P49708. Gene ID: 396054. Expression Region: His20~Cys164. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.