Learn More
Biomatik Corporation Recombinant Interferon Gamma (IFNg), 100UG
Supplier:  Biomatik Corporation RPU42142
Encompass_Preferred
Recombinant Mouse Interferon Gamma (IFNg) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interferon Gamma (IFNg). Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Accession Number: P01580. Gene ID: 15978. Expression Region: His23~Cys155. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5608