Learn More
Biomatik Corporation Recombinant Interferon Gamma Receptor 1 (IFNgR1), 100UG
Supplier:  Biomatik Corporation RPU42155
Encompass_Preferred
Recombinant Mouse Interferon Gamma Receptor 1 (IFNgR1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interferon Gamma Receptor 1 (IFNgR1). Target Synonyms: CD119. Accession Number: P15261. Gene ID: 15979. Expression Region: Gly23~Asp253. Tag Info: N-terminal GST Tag. Theoretical MW: 52kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5611