Learn More
Biomatik Corporation Recombinant Interleukin 1 Beta (IL1b), 100UG
Catalog No. 501505614
Encompass_Preferred
Recombinant Rat Interleukin 1 Beta (IL1b) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Interleukin 1 Beta (IL1b). Target Synonyms: IL1-B; IL1-Beta; IL1F2; IL-1β; Interleukin-1 Family Member 2; Catabolin. Accession Number: Q63264. Expression Region: Val117~Ser268. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5614 Supplier Biomatik Corporation Supplier No. RPU42174
Recombinant Rat Interleukin 1 Beta (IL1b) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Interleukin 1 Beta (IL1b). Target Synonyms: IL1-B; IL1-Beta; IL1F2; IL-1β; Interleukin-1 Family Member 2; Catabolin. Accession Number: Q63264. Expression Region: Val117~Ser268. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.