Learn More
Biomatik Corporation Recombinant Interleukin 24 (IL24), 100UG
Supplier:  Biomatik Corporation RPU42283
Encompass_Preferred
Recombinant Rat Interleukin 24 (IL24) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Interleukin 24 (IL24). Target Synonyms: C49A; FISP; IL10B; MDA7; Mob-5; ST16; Mda-7; Melanoma Differentiation Association Protein 7; Suppression Of Tumorigenicity 16. Accession Number: Q9JI24. Gene ID: 170819. Expression Region: Gln29~Leu183. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5617