Learn More
Biomatik Corporation Recombinant Interleukin 33 (IL33), 100UG
Supplier:  Biomatik Corporation RPU42296
Encompass_Preferred
Recombinant Mouse Interleukin 33 (IL33) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interleukin 33 (IL33). Target Synonyms: DV27; C9ORF26; IL1F11; NFHEV; Interleukin-1 Family, Member 11; Nuclear factor from high endothelial venules. Accession Number: Q8BVZ5. Gene ID: 77125. Expression Region: Ser109~Ile266. Tag Info: N-terminal GST Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5619