Learn More
Biomatik Corporation Recombinant Islet Cell Autoantigen 1 (ICA1), 100UG
Catalog No. 501505624
Encompass_Preferred
Recombinant Human Islet Cell Autoantigen 1 (ICA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Islet Cell Autoantigen 1 (ICA1). Target Synonyms: ICA69; ICAp69; ICA-p69; 69 kDa islet cell autoantigen. Accession Number: Q05084. Gene ID: 3382. Expression Region: Met1~Gly258. Tag Info: N-terminal His Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5624 Supplier Biomatik Corporation Supplier No. RPU42337
Recombinant Human Islet Cell Autoantigen 1 (ICA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Islet Cell Autoantigen 1 (ICA1). Target Synonyms: ICA69; ICAp69; ICA-p69; 69 kDa islet cell autoantigen. Accession Number: Q05084. Gene ID: 3382. Expression Region: Met1~Gly258. Tag Info: N-terminal His Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.