Learn More
Biomatik Corporation Recombinant Isocitrate Dehydrogenase 1, Soluble (IDH1), 100UG
Catalog No. 501505625
Encompass_Preferred
Recombinant Human Isocitrate Dehydrogenase 1, Soluble (IDH1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Isocitrate Dehydrogenase 1, Soluble (IDH1). Target Synonyms: PICD; IDP; Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; Cytosolic NADP-isocitrate dehydrogenase. Accession Number: O75874. Gene ID: 3417. Expression Region: Ala74~Ile333. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5625 Supplier Biomatik Corporation Supplier No. RPU42339
Add to Cart
Add to Cart
Recombinant Human Isocitrate Dehydrogenase 1, Soluble (IDH1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Isocitrate Dehydrogenase 1, Soluble (IDH1). Target Synonyms: PICD; IDP; Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; Cytosolic NADP-isocitrate dehydrogenase. Accession Number: O75874. Gene ID: 3417. Expression Region: Ala74~Ile333. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.