Learn More
Biomatik Corporation Recombinant Kallikrein 1 (KLK1), 100UG
Supplier:  Biomatik Corporation RPU42357
Encompass_Preferred
Recombinant Rat Kallikrein 1 (KLK1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Kallikrein 1 (KLK1). Target Synonyms: KLKR; Klk6; HK1; Kallikrein 1,Renal/Pancreas/Salivary; Kidney/pancreas/salivary gland kallikrein; Tissue kallikrein. Accession Number: P00758. Gene ID: 24594. Expression Region: Val21~Pro261. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5626