Learn More
Biomatik Corporation Recombinant Kallikrein 7 (KLK7), 100UG
Supplier:  Biomatik Corporation RPU42367
Encompass_Preferred
Recombinant Rat Kallikrein 7 (KLK7) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Kallikrein 7 (KLK7). Target Synonyms: PRSS6; SCCE; hK7; Kallikrein-Related Peptidase 7; Serine protease 6; Stratum corneum chymotryptic enzyme. Accession Number: P36373. Gene ID: 24523. Expression Region: Arg24~Glu255. Tag Info: N-terminal His and GST Tag. Theoretical MW: 55kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5627