Learn More
Biomatik Corporation Recombinant Dog Interleukin-18 (IL18), 100UG
Catalog No. 501504639
Encompass_Preferred
Recombinant Dog Interleukin-18 (IL18) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dog (Canis familiaris; Canine). Target Name: IL18. Target Synonyms: Interferon gamma-inducing factor; IFN-gamma-inducing factorInterleukin-1 gamma; IL-1 gamma. Accession Number: Q9XSR0. Expression Region: 37~193aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 22kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4639 Supplier Biomatik Corporation Supplier No. RPC20171
Recombinant Dog Interleukin-18 (IL18) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Dog (Canis familiaris; Canine). Target Name: IL18. Target Synonyms: Interferon gamma-inducing factor; IFN-gamma-inducing factorInterleukin-1 gamma; IL-1 gamma. Accession Number: Q9XSR0. Expression Region: 37~193aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 22kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.