Learn More
Biomatik Corporation Recombinant Keratin 18 (KRT18), 100UG
Supplier:  Biomatik Corporation RPU42385
Encompass_Preferred
Recombinant Human Cytokeratin 18 (CK18) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytokeratin 18 (CK18). Target Synonyms: KRT18; CYK18; K18; Keratin 18; Cell proliferation-inducing gene 46 protein; Keratin, type I cytoskeletal 18. Accession Number: P05783. Gene ID: 3875. Expression Region: Asp238~Leu396. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5629