Learn More
Biomatik Corporation Recombinant Kidney Injury Molecule 1 (Kim1), 100UG
Supplier:  Biomatik Corporation RPU42417
Encompass_Preferred
Recombinant Dog Kidney Injury Molecule 1 (Kim1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Dog (Canis familiaris; Canine). Target Name: Kidney Injury Molecule 1 (Kim1). Target Synonyms: HAVCR1; TIM1; TIMD1; HAVCR; Hepatitis A Virus Cellular Receptor 1; T Cell Immunoglobulin And Mucin Domain-Containing Protein 1; T-cell immunoglobulin mucin receptor 1. Accession Number: F1CME9. Gene ID: 612069. Expression Region: Gln23~Glu125. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5630