Learn More
Biomatik Corporation Recombinant Klotho (KL), 100UG
Catalog No. 501505633
Encompass_Preferred
Recombinant Mouse Klotho (KL) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Klotho (KL). Target Synonyms: KLa; Klotho Alpha. Accession Number: O35082. Gene ID: 16591. Expression Region: Leu59~Phe508. Tag Info: N-terminal His Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5633 Supplier Biomatik Corporation Supplier No. RPU42434
Recombinant Mouse Klotho (KL) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Klotho (KL). Target Synonyms: KLa; Klotho Alpha. Accession Number: O35082. Gene ID: 16591. Expression Region: Leu59~Phe508. Tag Info: N-terminal His Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.