Learn More
Biomatik Corporation Recombinant Lactate Dehydrogenase A (LDHA), 100UG
Catalog No. 501505635
Encompass_Preferred
Recombinant Mouse Lactate Dehydrogenase A (LDHA) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lactate Dehydrogenase A (LDHA). Target Synonyms: LDH-A; LDH-M; LDH1; PIG19; Cell proliferation-inducing gene 19 protein; LDH muscle subunit; Renal carcinoma antigen NY-REN-59. Accession Number: P06151. Gene ID: 16828. Expression Region: Met1~Phe332. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5635 Supplier Biomatik Corporation Supplier No. RPU42445
Recombinant Mouse Lactate Dehydrogenase A (LDHA) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lactate Dehydrogenase A (LDHA). Target Synonyms: LDH-A; LDH-M; LDH1; PIG19; Cell proliferation-inducing gene 19 protein; LDH muscle subunit; Renal carcinoma antigen NY-REN-59. Accession Number: P06151. Gene ID: 16828. Expression Region: Met1~Phe332. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.