Learn More
Biomatik Corporation Recombinant Lactoferrin (LTF), 100UG
Supplier:  Biomatik Corporation RPU42449
Encompass_Preferred
Recombinant Dog Lactoferrin (LTF) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Dog (Canis familiaris; Canine). Target Name: Lactoferrin (LTF). Target Synonyms: HLF2; LF; GIG12; Lactotransferrin; Growth-inhibiting protein 12; Talalactoferrin; Lfcin-H; Kaliocin-1. Accession Number: A0A8I3NMR3. Gene ID: 484787. Expression Region: Cys28~Lys351. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5636