Learn More
Biomatik Corporation Recombinant Laminin Alpha 4 (LAMa4), 100UG
Supplier:  Biomatik Corporation RPU42467
Encompass_Preferred
Recombinant Human Laminin Alpha 4 (LAMa4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Laminin Alpha 4 (LAMa4). Target Synonyms: LAM-A4; LAMA3. Accession Number: Q16363. Gene ID: 3910. Expression Region: Ile832~Lys1039. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5638