Learn More
Biomatik Corporation Recombinant Human Calpain-2 catalytic subunit (CAPN2), 100UG
Catalog No. 501504541
Encompass_Preferred
Recombinant Human Calpain-2 catalytic subunit (CAPN2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CAPN2. Target Synonyms: Calcium-activated neutral proteinase 2; CANP 2; Calpain M-typeCalpain large polypeptide L2; Calpain-2 large subunitMillimolar-calpain; M-calpain. Accession Number: P17655. Expression Region: 20~700aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 82.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4541 Supplier Biomatik Corporation Supplier No. RPC20016
Recombinant Human Calpain-2 catalytic subunit (CAPN2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CAPN2. Target Synonyms: Calcium-activated neutral proteinase 2; CANP 2; Calpain M-typeCalpain large polypeptide L2; Calpain-2 large subunitMillimolar-calpain; M-calpain. Accession Number: P17655. Expression Region: 20~700aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 82.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.