Learn More
Biomatik Corporation Recombinant Laminin Alpha 5 (LAMa5), 100UG
Supplier:  Biomatik Corporation RPU42471
Encompass_Preferred
Recombinant Mouse Laminin Alpha 5 (LAMa5) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Laminin Alpha 5 (LAMa5). Target Synonyms: LAM-A5. Accession Number: Q61001. Gene ID: 16776. Expression Region: Glu3517~His3718. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5639