Learn More
Biomatik Corporation Recombinant Laminin Beta 2 (LAMb2), 100UG
Supplier:  Biomatik Corporation RPU42473
Encompass_Preferred
Recombinant Mouse Laminin Beta 2 (LAMb2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Laminin Beta 2 (LAMb2). Target Synonyms: LAM-B2; LAMS; Laminin S; Laminin B1s chain; S-laminin subunit beta. Accession Number: Q61292. Gene ID: 16779. Expression Region: Gln1176~Asn1309. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5640