Learn More
Biomatik Corporation Recombinant Lecithin Cholesterol Acyltransferase (LCAT), 100UG
Supplier:  Biomatik Corporation RPU42492
Encompass_Preferred
Recombinant Mouse Lecithin Cholesterol Acyltransferase (LCAT) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lecithin Cholesterol Acyltransferase (LCAT). Target Synonyms: phosphatidylcholine-sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase. Accession Number: P16301. Gene ID: 16816. Expression Region: Ile290~Pro432. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5641