Learn More
Biomatik Corporation Recombinant Leukemia Inhibitory Factor (LIF), 100UG
Supplier:  Biomatik Corporation RPU42523
Encompass_Preferred
Recombinant Mouse Leukemia Inhibitory Factor (LIF) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Leukemia Inhibitory Factor (LIF). Target Synonyms: CDF; D-FACTOR; HILDA; MLPLI; Cholinergic Differentiation Factor; Differentiation-Stimulating Factor; Melanoma-Derived LPL Inhibitor; Emfilermin. Accession Number: P09056. Gene ID: 16878. Expression Region: Pro25~Phe203. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5642