Learn More
Biomatik Corporation Recombinant Lipopolysaccharide Binding Protein (LBP), 100UG
Catalog No. 501505644
Encompass_Preferred
Recombinant Human Lipopolysaccharide Binding Protein (LBP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lipopolysaccharide Binding Protein (LBP). Target Synonyms: LPS-Binding Protein. Accession Number: P18428. Gene ID: 3929. Expression Region: Asn27~Val481. Tag Info: N-terminal His Tag. Theoretical MW: 52kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5644 Supplier Biomatik Corporation Supplier No. RPU42568
Recombinant Human Lipopolysaccharide Binding Protein (LBP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lipopolysaccharide Binding Protein (LBP). Target Synonyms: LPS-Binding Protein. Accession Number: P18428. Gene ID: 3929. Expression Region: Asn27~Val481. Tag Info: N-terminal His Tag. Theoretical MW: 52kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.