Learn More
Biomatik Corporation Recombinant Lysophosphatidylcholine Acyltransferase 3 (LPCAT3), 100UG
Catalog No. 501505647
Encompass_Preferred
Recombinant Human Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysophosphatidylcholine Acyltransferase 3 (LPCAT3). Target Synonyms: MBOAT5; C3F; OACT5; Nessy; LPSAT; Membrane Bound O-AcylTransferase Domain Containing 5; Lysophospholipid Acyltransferase 5; Lysophosphatidylserine acyltransferase. Accession Number: Q6P1A2. Gene ID: 10162. Expression Region: Met122~Arg233. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5647 Supplier Biomatik Corporation Supplier No. RPU42611
Recombinant Human Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysophosphatidylcholine Acyltransferase 3 (LPCAT3). Target Synonyms: MBOAT5; C3F; OACT5; Nessy; LPSAT; Membrane Bound O-AcylTransferase Domain Containing 5; Lysophospholipid Acyltransferase 5; Lysophosphatidylserine acyltransferase. Accession Number: Q6P1A2. Gene ID: 10162. Expression Region: Met122~Arg233. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.