Learn More
Biomatik Corporation Recombinant Lysosomal Associated Membrane Protein 2 (LAMP2), 100UG
Supplier:  Biomatik Corporation RPU42615
Encompass_Preferred
Recombinant Human Lysosomal Associated Membrane Protein 2 (LAMP2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysosomal Associated Membrane Protein 2 (LAMP2). Target Synonyms: CD107-B; CD107b; LAMP-2C; LAMPB; LGP96; Lysosome-associated membrane glycoprotein 2; CD107 antigen-like family member B. Accession Number: P13473. Gene ID: 3920. Expression Region: Lys193~Asn375. Tag Info: N-terminal His and GST Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5648