Learn More
Biomatik Corporation Recombinant Human Myosin-9 (MYH9), partial, 100UG
Supplier:  Biomatik Corporation RPC20174
Encompass_Preferred
Recombinant Human Myosin-9 (MYH9), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MYH9. Target Synonyms: Cellular myosin heavy chain, type AMyosin heavy chain 9Myosin heavy chain, non-muscle IIaNon-muscle myosin heavy chain A; NMMHC-ANon-muscle myosin heavy chain IIa; NMMHC II-a; NMMHC-IIA. Accession Number: P35579. Expression Region: 2~241aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 54.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4641