Learn More
Biomatik Corporation Recombinant Lysyl Oxidase (LOX), 100UG
Supplier:  Biomatik Corporation RPU42622
Encompass_Preferred
Recombinant Human Lysyl Oxidase (LOX) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysyl Oxidase (LOX). Target Synonyms: Protein-Lysine 6-Oxidase. Accession Number: P28300. Gene ID: 4015. Expression Region: Pro213~Tyr417. Tag Info: N-terminal His and GST Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5649