Learn More
Biomatik Corporation Recombinant Lysyl Oxidase (LOX), 100UG
Catalog No. 501505650
Encompass_Preferred
Recombinant Mouse Lysyl Oxidase (LOX) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lysyl Oxidase (LOX). Target Synonyms: Protein-Lysine 6-Oxidase. Accession Number: P28301. Gene ID: 16948. Expression Region: Pro207~Tyr411. Tag Info: N-terminal His and GST Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5650 Supplier Biomatik Corporation Supplier No. RPU42623
Recombinant Mouse Lysyl Oxidase (LOX) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lysyl Oxidase (LOX). Target Synonyms: Protein-Lysine 6-Oxidase. Accession Number: P28301. Gene ID: 16948. Expression Region: Pro207~Tyr411. Tag Info: N-terminal His and GST Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.