Learn More
Biomatik Corporation Recombinant Lysyl Oxidase Like Protein 1 (LOXL1), 100UG
Supplier:  Biomatik Corporation RPU42624
Encompass_Preferred
Recombinant Human Lysyl Oxidase Like Protein 1 (LOXL1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysyl Oxidase Like Protein 1 (LOXL1). Target Synonyms: LOL; LOXL; Lysyl oxidase homolog 1. Accession Number: Q08397. Gene ID: 4016. Expression Region: Leu369~Ser574. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5651