Learn More
Biomatik Corporation Recombinant Major Basic Protein (MBP), 100UG
Supplier:  Biomatik Corporation RPU42660
Encompass_Preferred
Recombinant Human Major Basic Protein (MBP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Major Basic Protein (MBP). Target Synonyms: PRG2; BMPG; Proteoglycan 2; Bone Marrow,Natural Killer Cell Activator; Eosinophil Granule Major Basic Protein; Proteoglycan 2,Bone Marrow; Pregnancy-associated major basic. Accession Number: P13727. Gene ID: 5553. Expression Region: Gln105~Tyr222. Tag Info: N-terminal His and GST Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5652