Learn More
Biomatik Corporation Recombinant Major Histocompatibility Complex Class I E (MHCE), 100UG
Supplier:  Biomatik Corporation RPU42664
Encompass_Preferred
Recombinant Human Major Histocompatibility Complex Class I E (MHCE) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Major Histocompatibility Complex Class I E (MHCE). Target Synonyms: HLAE; EA1.2; EA2.1; HLA-6.2; MHC-E; HLA-E; Leukocyte Antigen E; MHC class I antigen E; HLA class I histocompatibility antigen, alpha chain E. Accession Number: P13747. Gene ID: 3133. Expression Region: Gly22~Leu358. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5653