Learn More
Biomatik Corporation Recombinant Major Histocompatibility Complex Class I G (MHCG), 100UG
Supplier:  Biomatik Corporation RPU42665
Encompass_Preferred
Recombinant Human Major Histocompatibility Complex Class I G (MHCG) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Major Histocompatibility Complex Class I G (MHCG). Target Synonyms: HLA-G; MHC-G; HLAG; Leukocyte Antigen G; b2 microglobulin; HLA class I histocompatibility antigen, alpha chain G. Accession Number: P17693. Gene ID: 3135. Expression Region: Met29~His117. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5654