Learn More
Biomatik Corporation Recombinant Matrix Metalloproteinase 7 (MMP7), 100UG
Supplier:  Biomatik Corporation RPU42715
Encompass_Preferred
Recombinant Human Matrix Metalloproteinase 7 (MMP7) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Matrix Metalloproteinase 7 (MMP7). Target Synonyms: MPSL1; MAT; PUMP-1; Matrilysin Uterine; Matrilysin; Matrin; Pump-1 Protease; Uterine Metalloproteinase. Accession Number: P09237. Gene ID: 4316. Expression Region: Met1~Lys267. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5658