Learn More
Biomatik Corporation Recombinant Microtubule Associated Protein 2 (MAP2), 100UG
Catalog No. 501505662
Encompass_Preferred
Recombinant Human Microtubule Associated Protein 2 (MAP2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Microtubule Associated Protein 2 (MAP2). Target Synonyms: MAP2A; MAP2B; MAP2C. Accession Number: P11137. Gene ID: 4133. Expression Region: Leu727~Asp985. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5662 Supplier Biomatik Corporation Supplier No. RPU42754
Recombinant Human Microtubule Associated Protein 2 (MAP2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Microtubule Associated Protein 2 (MAP2). Target Synonyms: MAP2A; MAP2B; MAP2C. Accession Number: P11137. Gene ID: 4133. Expression Region: Leu727~Asp985. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.