Learn More
Biomatik Corporation Recombinant Midkine (MK), 100UG
Supplier:  Biomatik Corporation RPU42760
Encompass_Preferred
Recombinant Rat Midkine (MK) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Midkine (MK). Target Synonyms: MDK; ARAP; NEGF2; Neurite Growth-Promoting Factor 2; Amphiregulin-associated protein; Midgestation and kidney protein; Neurite outgrowth-promoting factor 2. Accession Number: Q9R1S9. Gene ID: 81517. Expression Region: Ala22~Asp140. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5663