Learn More
Biomatik Corporation Recombinant Mucin 1 (MUC1), 100UG
Catalog No. 501505665
Encompass_Preferred
Recombinant Human Mucin 1 (MUC1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mucin 1 (MUC1). Target Synonyms: CD227; EMA; H23AG; KL-6; PEM; PEMT; PUM; CA15-3; CA153; Cancer antigen 15-3; Carcinoma-associated mucin; Episialin; Peanut-reactive urinary mucin; Polymorphic epithelial mucin. Accession Number: P15941. Gene ID: 4582. Expression Region: Ser1098~Leu1255. Tag Info: N-terminal His and GST Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5665 Supplier Biomatik Corporation Supplier No. RPU42815
Recombinant Human Mucin 1 (MUC1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mucin 1 (MUC1). Target Synonyms: CD227; EMA; H23AG; KL-6; PEM; PEMT; PUM; CA15-3; CA153; Cancer antigen 15-3; Carcinoma-associated mucin; Episialin; Peanut-reactive urinary mucin; Polymorphic epithelial mucin. Accession Number: P15941. Gene ID: 4582. Expression Region: Ser1098~Leu1255. Tag Info: N-terminal His and GST Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.