Learn More
Biomatik Corporation Recombinant Mucin 2 (MUC2), 100UG
Supplier:  Biomatik Corporation RPU42821
Encompass_Preferred
Recombinant Human Mucin 2 (MUC2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mucin 2 (MUC2). Target Synonyms: MLP; SMUC; Oligomeric Mucus/Gel-Forming; Mucin 2,Intestinal/Tracheal. Accession Number: Q02817. Gene ID: 4583. Expression Region: Cys5075~Gly5179. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5666