Learn More
Biomatik Corporation Recombinant Myosin Heavy Chain 6, Cardiac Muscle, Alpha (MYH6), 100UG
Supplier:  Biomatik Corporation RPU42877
Encompass_Preferred
Recombinant Rat Myosin Heavy Chain 6, Cardiac Muscle, Alpha (MYH6) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Myosin Heavy Chain 6, Cardiac Muscle, Alpha (MYH6). Target Synonyms: ASD3; MYHC; MYHCA; MyHC-alpha; Cardiomyopathy,Hypertrophic 1; Myosin heavy chain, cardiac muscle alpha isoform. Accession Number: P02563. Expression Region: Gln1747~Glu1938. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5671