Learn More
Biomatik Corporation Recombinant Neprilysin (NEP), 100UG
Supplier:  Biomatik Corporation RPU42988
Encompass_Preferred
Recombinant Rat Neprilysin (CD10) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Neprilysin (CD10). Target Synonyms: NEP; MME; CALLA; SFE; Atriopeptidase; Enkephalinase; Skin fibroblast elastase; Common Acute Lymphoblastic Leukemia Antigen; Membrane Metallo-Endopeptidase. Accession Number: P07861. Gene ID: 24590. Expression Region: Ala286~Val428. Tag Info: N-terminal GST Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5673