Learn More
Biomatik Corporation Recombinant Neuregulin 1 (NRG1), 100UG
Supplier:  Biomatik Corporation RPU43000
Encompass_Preferred
Recombinant Mouse Neuregulin 1 (NRG1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Neuregulin 1 (NRG1). Target Synonyms: ARIA; GGF; HGL; HRG; HRGA; NDF; SMDF; Acetylcholine receptor-inducing activity; Glial growth factor; Heregulin; Neu differentiation factor; Sensory and motor neuron-derived factor. Accession Number: Q6DR98. Expression Region: Ser166~His280. Tag Info: N-terminal His and GST Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5675