Learn More
Biomatik Corporation Recombinant Non Metastatic Cells 2, Protein NM23B Expressed In (NME2), 100UG
Supplier:  Biomatik Corporation RPU43087
Encompass_Preferred
Recombinant Mouse Non Metastatic Cells 2, Protein NM23B Expressed In (NME2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Non Metastatic Cells 2, Protein NM23B Expressed In (NME2). Target Synonyms: NDPKB; NM23-H2; NM23B; Puf; Nucleoside diphosphate kinase B; C-myc purine-binding transcription factor PUF; Histidine protein kinase NDKB. Accession Number: Q01768. Gene ID: 18103. Expression Region: Phe8~Trp149. Tag Info: N-terminal His Tag. Theoretical MW: 23/25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5679