Learn More
Biomatik Corporation Recombinant Nuclear Factor Kappa B (NFkB), 100UG
Supplier:  Biomatik Corporation RPU43097
Encompass_Preferred
Recombinant Human Nuclear Factor Kappa B (NFkB) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Nuclear Factor Kappa B (NFkB). Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Accession Number: P19838. Gene ID: 4790. Expression Region: Pro42~Met367. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5680