Learn More
Biomatik Corporation Recombinant Nuclear Receptor Subfamily 3, Group C, Member 1 (NR3C1), 100UG
Supplier:  Biomatik Corporation RPU43103
Encompass_Preferred
Recombinant Human Glucocorticoid Receptor (GR) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glucocorticoid Receptor (GR). Target Synonyms: GCCR; GCR; NR3C1; GRL; NR3-C1; Nuclear Receptor Subfamily 3, Group C, Member 1. Accession Number: P04150. Gene ID: 2908. Expression Region: Leu532~Glu730. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5681