Learn More
Biomatik Corporation Recombinant Nucleophosmin (NPM), 100UG
Supplier:  Biomatik Corporation RPU43111
Encompass_Preferred
Recombinant Mouse Nucleophosmin 1 (NPM1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Nucleophosmin 1 (NPM1). Target Synonyms: B23; NPM; Nucleolar Phosphoprotein B23; Numatrin; Nucleophosmin/Nucleoplasmin Family,Member 1; Nucleolar protein NO38. Accession Number: Q61937. Gene ID: 18148. Expression Region: Gly20~Lys154. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5683