Learn More
Biomatik Corporation Recombinant Nucleoporin 98kDa (NUP98), 100UG
Supplier:  Biomatik Corporation RPU43125
Encompass_Preferred
Recombinant Human Nucleoporin 98 (NUP98) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Nucleoporin 98 (NUP98). Target Synonyms: ADIR2; NUP96; Nucleoporin 98kDa. Accession Number: P52948. Gene ID: 4928. Expression Region: Gln629~Pro894. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5684